missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Nutrient Broth

Catalog No. DF0003178
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0003178 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0003178 Supplier BD Supplier No. 234000
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

General purpose medium for the cultivation of microorganisms with nonexacting nutritional requirements

Specifications

Description Nutrient Broth
Form Dehydrated
For Use With (Application) General purpose medium for the cultivation of microorganisms with non-exacting nutritional requirements
Quantity 500 g