missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: TT (Tetrathionate) Broth Base, Hajna

Catalog No. DF0491177
Change view
Click to view available options
Quantity:
500 g
Packaging Type:
Bottle
Media Type:
TT Broth Base, Hajna
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type Media Type
DF0491177 500 g Bottle TT Broth Base, Hajna
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0491177 Supplier BD Supplier No. 249120
Edge

Selective enrichment for Salmonella from food and dairy products

Specifications

Certifications/Compliance USDA
Content And Storage Refrigerate
Form Dehydrated
Media Type TT Broth Base, Hajna
Product Type Microbiology Media
Formulation Yeast extract 2g/L, Tryptose 18g/L, Dextrose 0.5g/L, D-Mannitol 2.5g/L, Sodium Desoxycholate 0.5g/L, Sodium Chloride 5g/L, Sodium Thiosulfate 38g/L, Calcium Carbonate 25g/L, Brilliant green 0.01g/L
For Use With (Application) For enriching Salmonella from food and dairy products prior to isolation procedures
Packaging Type Bottle
Quantity 500 g