missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Tryptic Soy Agar (Soybean-Casein Digest Agar)

Catalog No. DF0369078
Change view
Click to view available options
Quantity:
500 g
2 kg
10 kg
Packaging Type:
Bottle
Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
DF0369078 2 kg Pail
DF0369176 500 g Bottle
DF0369087 10 kg Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. DF0369078 Supplier BD Supplier No. 236920
Only null left

Use dehydrated media for isolation, detection and cultivation of a wide variety of microorganisms. BD Difco™ Dehydrated Culture Media: Tryptic Soy Agar (Soybean-Casein Digest Agar) is supplied in dehydrated form for the convenience of the laboratorian and bioprocessing specialist.

Applications:

  • Used with or without blood for cultivation and isolation of variety of organisms
  • Used for sterility testing

Certifications

AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP

Specifications

Certifications/Compliance AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP
Content And Storage 2°C to 30°C
Form Dehydrated
Media Type Tryptic Soy Agar
Product Type Tryptic Soy Agar
Formulation Pancreatic digest of Casein 15g/L, Papaic digest of Soybean 5g/L, Sodium Chloride 5g/L, Agar 15g/L
For Use With (Application) Isolation and cultivation of non-fastidious and fastidious microorganisms
Packaging Type Pail
Quantity 2 kg