missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Triple Sugar Iron Agar
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, ISO, MCM9, SMD, SMWW, USDA |
| Form | Dehydrated |
| Media Type | Triple Sugar Iron Agar |
| Product Type | Agar |
| Formulation | Beef extract 3g/L, Yeast extract 3g/L, Pancreatic digest of Casein 15g/L, Proteose Peptone No. 3 5g/L, Dextrose 1g/L, Lactose 10g/L, Sucrose 10g/L, Ferrous Sulfate 0.2g/L, Sodium Chloride 5g/L, Sodium Thiosulfate 0.3g/L, Agar 12g/L, Phenol red 24mg/L |
| For Use With (Application) | For microbial and molecular Genetics |
| Packaging Type | Bottle |
| Quantity | 500 g |