missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Potato Dextrose Agar

For cultivation of Yeasts and molds. BD Difco™ Dehydrated Culture Media: Potato Dextrose Agar is supplied in dehydrated format.

$242.82 - $884.71

Specifications

Certifications/Compliance ISO/DIS, 11014, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, JP, MCM9, SMD, USP
Content And Storage 2°C to 30°C
Form Dehydrated
Media Type Potato Dextrose Agar
Product Type Agar
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Type Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Type Price Quantity  
DF0013-07-8
View Documents
BD
213200
2000 g Pail
Each for $884.71
Only null left
 
DF0013-17-6
View Documents
BD
213400
500 g Bottle
Each for $242.82
Only null left
 
Description

Description

  • Dehydrated format
  • Culture media
  • DNR (Latex) free
Specifications

Specifications

ISO/DIS, 11014, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, JP, MCM9, SMD, USP
Dehydrated
Agar
2°C to 30°C
Potato Dextrose Agar
Isolation of Yeasts and molds
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'