missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Potato Dextrose Agar
Description
- Dehydrated format
- Culture media
- DNR (Latex) free
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, JP, MCM9, SMD, USP |
| Content And Storage | 2°C to 30°C |
| Form | Dehydrated |
| Media Type | Potato Dextrose Agar |
| Product Type | Agar |
| Formulation | Potato Starch (from infusion) 4g/L, Dextrose 20g/L, Agar 15g/L |
| For Use With (Application) | Isolation of Yeasts and molds |
| Packaging Type | Bottle |
| Quantity | 500 g |