missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Potato Dextrose Agar

Catalog No. DF0013078
Change view
Click to view available options
Quantity:
500 g
2000 g
Packaging Type:
Bottle
Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
DF0013078 2000 g Pail
DF0013176 500 g Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. DF0013078 Supplier BD Supplier No. 213200

For cultivation of Yeasts and molds. BD Difco™ Dehydrated Culture Media: Potato Dextrose Agar is supplied in dehydrated format.

  • Dehydrated format
  • Culture media
  • DNR (Latex) free

Specifications

Certifications/Compliance ISO/DIS, 11014, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, JP, MCM9, SMD, USP
Content And Storage 2°C to 30°C
Form Dehydrated
Media Type Potato Dextrose Agar
Product Type Agar
Formulation Potato Starch (from infusion) 4g/L, Dextrose 20g/L
For Use With (Application) Isolation of Yeasts and molds
Packaging Type Pail
Quantity 2000 g