missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Membrane Fecal Coliform (mFC) Agar

Catalog No. DF0677173
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0677173 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0677173 Supplier BD Supplier No. 267720
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For detection and enumeration of fecal coliforms by the membrane filter technique at elevated temperatures

  • Use with Rosolic Acid
  • Formulation: Tryptose 10g/L, Proteose Peptone No. 3 5g/L, Yeast extract 3g/L, Lactose 12.5g/L, Bile Salts No. 3 1.5g/L, Sodium Chloride 5g/L, Agar 15g/L, Aniline blue 0.1g/L

Certifications

AOAC, CCAM, EPA, SMWW

Specifications

Certifications/Compliance AOAC, CCAM, EPA, SMWW
Content And Storage Refrigerate
Form Dehydrated
Media Type mFC Agar
Product Type Nutrient Agar
Formulation Tryptose 10g/L, Proteose Peptone No. 3 5g/L, Yeast extract 3g/L, Lactose 12.5g/L, Bile Salts No. 3 1.5g/L, Sodium Chloride 5g/L, Agar 15g/L, Aniline blue 0.1g/L
For Use With (Application) For Rosolic Acid in cultivating and enumerating fecal Coliforms by the membrane filter technique at elevated temperatures
Packaging Type Bottle
Quantity 500 g