missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: MacConkey Agar

Catalog No. DF0075073
Change view
Click to view available options
Quantity:
2 kg
Packaging Type:
Pail
Certifications/Compliance:
CE, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, JP, MCM9, SMD, SMWW, USP
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type Certifications/Compliance
DF0075073 2 kg Pail CE, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, JP, MCM9, SMD, SMWW, USP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0075073 Supplier BD Supplier No. 212122
Only null left

For differential isolation of enteric bacilli based on lactose fermentation

Specifications

Certifications/Compliance CE, AOAC, BAM, BS12, CCAM, CMPH2, COMPF, EP, JP, MCM9, SMD, SMWW, USP
Content And Storage Refrigerate
Form Dehydrated
Media Type MacConkey Agar
Product Type Agar
Formulation Pancreatic digest of Gelatin 17g/L, Peptones (meat and Casein) 3g/L, Lactose 10g/L, Bile Salts 1.5g/L, Sodium Chloride 5g/L, Agar 13.5g/L, Neutral red 0.03g/L, Crystal Violet 1mg/L
For Use With (Application) For differential isolation of enteric Bacilli based on Lactose fermentation
Packaging Type Pail
Quantity 2 kg