missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Hektoen Enteric Agar

Catalog No. DF0853179
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0853179 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0853179 Supplier BD Supplier No. 285340
Edge

For selective, differential isolation of Salmonella and Shigella from other gram-negative enteric bacilli

Specifications

Certifications/Compliance AOAC, BAM, BS12, CCAM, CMPH2, COMPF, MCM9, SMD
Content And Storage <30°C
Form Dehydrated
Media Type Hektoen Enteric Agar
Product Type Nutrient Agar
Formulation Proteose Peptone 12g/L, Yeast extract 3g/L, Bile Salts No. 3 9g/L, Lactose 12g/L, Saccharose 12g/L, Salicin 2g/L, Sodium Chloride 5g/L, Sodium Thiosulfate 5g/L, Ferric Ammonium Citrate 1.5g/L, Agar 14g/L, Bromthymol blue 65mg/L, Acid Fuchsin 0.1g/L
For Use With (Application) Isolation and cultivation of gram negative enteric microorganisms, especially Shigella, from a variety of clinical and nonclinical specimens
Packaging Type Bottle
Quantity 500 g