missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Fluid Thioglycollate Medium

Catalog No. DF0256172
Change view
Click to view available options
Quantity:
500 g
2000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0256172 500 g
DF0256074 2000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. DF0256172 Supplier BD Supplier No. 225650
Edge

For sterility testing of biologics and cultivation of anaerobes, aerobes and microaerophiles

  • Formulation: Pancreatic digest of Casein 15g/L, Yeast extract 5g/L, Dextrose 5.5g/L, Sodium Chloride 2.5g/L, L-Cystine 0.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.75g/L, Resazurin 1mg/L

Certifications

AOAC, BAM, BS12, COMPF, EP, USDA, USP

Specifications

Certifications/Compliance AOAC, BAM, BS12, COMPF, EP, USDA, USP
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Fluid Thioglycollate Medium
Product Type Dehydrated Culture Media
Formulation Pancreatic digest of Casein 15g/L, Yeast extract 5g/L, Dextrose 5.5g/L, Sodium Chloride 2.5g/L, L-Cystine 0.5g/L, Sodium Thioglycollate 0.5g/L, Agar 0.75g/L, Resazurin 1mg/L
For Use With (Application) Used for sterility testing
Packaging Type Bottle
Quantity 500 g