missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Chromogenic Dehydrated Media: m-EI Agar Base

Catalog No. B14881
Change view
Click to view available options
Quantity:
500 g
Certifications/Compliance:
ISO/DIS, 11014
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Certifications/Compliance
B14881 500 g ISO/DIS, 11014
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B14881 Supplier BD Supplier No. 214881
Only null left

Culture media tests water for the presence of Enterococci by membrane filtration method

  • Any colonies present will exhibit blue halo within 24 hours of the test
  • Conforms to EPA Method 1600

Order Info

Available in dehydrated form.

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage 5°C to 25°C
Form Solid
Manufacturing Origin United States
Media Type m EI Agar
Formulation Peptone 10g/L, Sodium Chloride 15g/L, Esculin 1g/L, Cycloheximide 0.05g/L, Sodium Azide 0.15g/L, Yeast extract 30g/L, Indoxyl-beta-D-glucoside 0.75g/L, Agar 15g/L
For Use With (Application) The chromogenic detection and enumeration of enterococci in Water by the single-step membrane filtration technique
Packaging Type Bottle
Quantity 500 g