missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Chromogenic Dehydrated Culture Media: Modified mTEC Agar

Catalog No. b14880
Change view
Click to view available options
Quantity:
500 g
Certifications/Compliance:
ISO/DIS, 11014
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Certifications/Compliance
B14880 500 g ISO/DIS, 11014
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B14880 Supplier BD Supplier No. 214880
Only null left

Culture media tests water for the presence of E.coli with membrane filtration method

Detects degree of water quality deterioration in drinking distribution system enumerates, and monitors the progress in remediation. Provides precise and quantifiable results; also less time consuming and costly than other techniques.

  • Any colonies present will exhibit red or magenta color within 24 hours of the test
  • Conforms to EPA Method 1603

Order Info

Available in prepared 15 x 60mm plates.

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Beige
Content And Storage 2 to 25°C
Form Solid
Manufacturing Origin United States
Media Type Modified m TEC Agar
Formulation No. 3 Proteose Peptone 5g/L, Yeast extract 3g/L, Lactose 10g/L, Sodium Chloride 7.5g/L, Dipotassium Phosphate 3.3g/L, Monopotassium Phosphate 1g/L, Sodium Lauryl Sulfate 0.2g/L, Sodium Desoxycholate 0.1g/L, 5-Bromo-6-chloro-3-indolyl-beta-D-glucuronide 0.5g/L, Agar 15g/L
For Use With (Application) The chromogenic detection and enumeration of thermotolerant Escherichia coli in Water by the membrane filtration technique
Packaging Type Bottle
Quantity 500 g