missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Chromogenic Dehydrated Culture Media: MI Agar
Description
Detects degree of water quality deterioration in drinking distribution system enumerates, and monitors the progress in remediation. Provides precise and quantifiable results; also less time consuming and costly than other techniques.
- Chromogenic-fluorogenic medium for detecting total coliforms and Escherichia coli in 24 hours or less
- Under ambient light, E. coli exhibits bluish-gray appearance; under long-wave UV light, it displays blue-green fluorescence
- Coliform are cream under ambient light, and display blue-white fluorescence under long-wave UV light
- Conforms to EPA Method 1604 for monitoring drinking water under the Total Coliform Rule, and source water under the Surface Water Treatment
Applications:
For testing ambient and potable water, treatment plant effluents, and beverage and pharmaceutical manufacturing
Order Info
Available in dehydrated form.
Specifications
Specifications
| Certifications/Compliance | ISO, 9000 |
| Color | Beige |
| Content And Storage | 2 to 25°C |
| Form | Solid |
| Manufacturing Origin | United States |
| Media Type | MI Agar |
| Formulation | Proteose Peptone No. 3 5g/L, Yeast extract 3g/L, D-Lactose 1g/L, 4-Methylumbelliferyl-beta-D-galactopyranoside 0.1g/L, Indoxyl-beta-D-glucuronide 0.32g/L, Sodium Chloride 7.5g/L, Dipotassium Phosphate 3.3g/L, Monopotassium Phosphate 1g/L, Sodium Lauryl Sulfate 0.2g/L, Sodium Desoxycholate 0.1g/L, Agar 15g/L |
| For Use With (Application) | Detection and enumerate Escherichia coli and total Coliforms in drinking Water by the membrane filtration technique |
| Packaging Type | Plate |
| Quantity | 500 g |