missing translation for 'onlineSavingsMsg'
Learn More

Diethylphosphonoacetic acid, 98%

Catalog No. AC330670050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC330670050 5 g Glass bottle
AC330670250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC330670050 Supplier Thermo Scientific Chemicals Supplier No. 330670050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3095-95-2 | C6H13O5P | 196.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)

Chemical Identifiers

CAS 3095-95-2
Molecular Formula C6H13O5P
Molecular Weight (g/mol) 196.14
MDL Number MFCD00192032
InChI Key DVQMPWOLBFKUMM-UHFFFAOYSA-N
Synonym 2-diethoxyphosphoryl acetic acid, diethylphosphonoacetic acid, diethyl carboxymethylphosphonate, diethoxyphosphorylacetic acid, diethyl phosphonoacetate, diethoxyphosphoryl acetic acid, acetic acid, diethoxyphosphinyl, acmc-20al82, diethylphosphonoethanoic acid, diethylcarboxymethylphosphonate
PubChem CID 36704
IUPAC Name 2-diethoxyphosphorylacetic acid
SMILES CCOP(=O)(CC(=O)O)OCC

Specifications

Color Colorless to Yellow
Density 1.2200g/mL
Boiling Point 150.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4440 to 1.4460
Quantity 5 g
Specific Gravity 1.22
Formula Weight 196.14
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material Diethylphosphonoacetic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only