missing translation for 'onlineSavingsMsg'
Learn More

Diethylmethoxyborane, 1M soln in THF

Catalog No. AAL15107AC
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL15107AC 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL15107AC Supplier Thermo Scientific Chemicals Supplier No. L15107AC
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7397-46-8 | C5H13BO | 99.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7397-46-8
Molecular Formula C5H13BO
Molecular Weight (g/mol) 99.97
MDL Number MFCD00013240
InChI Key FESAXEDIWWXCNG-UHFFFAOYSA-N
Synonym methoxydiethylborane, methyl diethylborinate, diethyl methoxy borane, borinic acid, diethyl-, methyl ester, diethyl methoxyborane, unii-3ah40d7l25, diethyl-methoxyborane, methoxy-diethylborane, methoxydiethyl-borane
PubChem CID 522516
SMILES CCB(CC)OC

Specifications

Density 0.868
Flash Point −6°C (21°F)
Refractive Index 1.404
Linear Formula (CH3CH2)2BOCH3
Quantity 25 mL
UN Number UN2924
Beilstein 2231225
Sensitivity Air and moisture sensitive
IUPAC Name diethyl(methoxy)borane
Formula Weight 99.97
Concentration or Composition (by Analyte or Components) 1M soln in THF
Chemical Name or Material Diethylmethoxyborane
Show More Show Less
Hazard Category H225-H250-H302-H314-H317-H335-H336-H351-H373
Hazard Statement GHS H Statement
H225-H301-H314-H318
Highly flammable liquid and vapor.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P201-P202-P210-P222-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P272-P280g-P281-P302+P334-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
RTECSNumber ED6151000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only