missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution

Catalog No. ac407290010
Change view
Click to view available options
Quantity:
250 g
1 kg
3 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC407290010 1 kg Glass bottle
AC407292500 250 g Glass bottle
AC407290030 3 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC407290010 Supplier Thermo Scientific Chemicals Supplier No. 407290010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 140-01-2 | C14H18N3Na5O10 | 503.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent

Specifications

Chemical Name or Material Diethylenetriaminepentaacetic acid, pentasodium salt
Name Note 40% Aqueous Solution
CAS 140-01-2, 7732-18-5
Melting Point -40°C
Density 1.2900g/mL
pH 11.0 to 12.0 (1% aq. soln.)
Boiling Point 106°C
Flash Point >100°C
Packaging Glass bottle
Molecular Formula C14H18N3Na5O10
Refractive Index 1.4185 to 1.4205
Linear Formula NaOCOCH2N(CH2CH2N(CH2COONa)2)2
Quantity 1 kg
MDL Number MFCD00051016
Beilstein 04, III, 1190
Specific Gravity 1.29
Synonym pentasodium dtpa, tetralon b, trilon c, versenex 80, pentasodium pentetate, hamp-ex 80, detarex py, kiresuto p, chelest p, plexene d
Solubility Information Solubility in water: soluble. Other solubilities: mixible with many organic solvents
InChI Key LQPLDXQVILYOOL-UHFFFAOYSA-I
SMILES [Na+].[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CCN(CC([O-])=O)CC([O-])=O
IUPAC Name pentasodium;2-[bis[2-[bis(carboxylatomethyl)amino]ethyl]amino]acetate
Molecular Weight (g/mol) 503.26
PubChem CID 8779
Formula Weight 503.26
Percent Purity 39 to 41%
Grade Technical
Physical Form Solution
Show More Show Less

RUO – Research Use Only