missing translation for 'onlineSavingsMsg'
Learn More

Diethylenetriaminepentaacetic Acid Calcium Trisodium Salt Hydrate 98.0+%, TCI America™

Catalog No. D18695G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D18695G 5 g
D186925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D18695G Supplier TCI America Supplier No. D18695G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 207226-35-5
Molecular Formula C14H20CaN3Na3O11
Molecular Weight (g/mol) 515.37
MDL Number MFCD00150947
InChI Key AWBWYUNJDQLQGT-UHFFFAOYSA-I
Synonym Calcium Trisodium Diethylenetriaminepentaacetate
PubChem CID 17749130
IUPAC Name calcium trisodium 2-[bis({2-[bis(carboxylatomethyl)amino]ethyl})amino]acetate hydrate
SMILES O.[Na+].[Na+].[Na+].[Ca++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CCN(CC([O-])=O)CC([O-])=O

Specifications

Color White
Quantity 5 g
Formula Weight 497.36
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Diethylenetriaminepentaacetic Acid Calcium Trisodium Salt Hydrate
RTECSNumber MB8210000
TSCA Yes