Learn More
Diethylenetriaminepentaacetic acid, 98+%
CAS: 67-43-6 | C14H23N3O10 | 393.34 g/mol
Supplier: Thermo Scientific Chemicals 114320010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagentSpecifications
| Diethylenetriaminepentaacetic acid | |
| 220.0°C | |
| 1.5600g/mL | |
| Authentic | |
| Plastic bottle | |
| 1 kg | |
| 1.56 | |
| pentetic acid, diethylenetriaminepentaacetic acid, dtpa, detapac, complexon v, detarex, perma kleer, titriplex v, monaquest cai, hamp-ex acid | |
| QPCDCPDFJACHGM-UHFFFAOYSA-N | |
| 2-[bis[2-[bis(carboxymethyl)amino]ethyl]amino]acetic acid | |
| 3053 | |
| 393.34 | |
| Crystalline Powder |
| 67-43-6 | |
| White | |
| 200°C | |
| 98+% | |
| C14H23N3O10 | |
| 04,III,119 | |
| 15,7237 | |
| Solubility in water: 5g/L (20°C), 13g/L (50°C), 48g/L (80°C). Other solubilities: soluble in alkali | |
| C(CN(CC(=O)O)CC(=O)O)N(CCN(CC(=O)O)CC(=O)O)CC(=O)O | |
| 393.34 | |
| CHEBI:35739 | |
| 98+% |
Chemical Identifiers
| 67-43-6 | |
| 393.34 | |
| pentetic acid, diethylenetriaminepentaacetic acid, dtpa, detapac, complexon v, detarex, perma kleer, titriplex v, monaquest cai, hamp-ex acid | |
| CHEBI:35739 | |
| C(CN(CC(=O)O)CC(=O)O)N(CCN(CC(=O)O)CC(=O)O)CC(=O)O |
| C14H23N3O10 | |
| QPCDCPDFJACHGM-UHFFFAOYSA-N | |
| 3053 | |
| 2-[bis[2-[bis(carboxymethyl)amino]ethyl]amino]acetic acid |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 200-652-8
RUO – Research Use Only