missing translation for 'onlineSavingsMsg'
Learn More

Diethylenetriaminepentaacetic acid, 98+%

Catalog No. AC114322500
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC114322500 250 g Plastic bottle
AC114320010 1 kg Plastic bottle
AC114320050 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC114322500 Supplier Thermo Scientific Chemicals Supplier No. 114322500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 67-43-6 | C14H23N3O10 | 393.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent

Chemical Identifiers

CAS 67-43-6
Molecular Formula C14H23N3O10
Molecular Weight (g/mol) 393.34
InChI Key QPCDCPDFJACHGM-UHFFFAOYSA-N
Synonym pentetic acid, diethylenetriaminepentaacetic acid, dtpa, detapac, complexon v, detarex, perma kleer, titriplex v, monaquest cai, hamp-ex acid
PubChem CID 3053
ChEBI CHEBI:35739
IUPAC Name 2-[bis[2-[bis(carboxymethyl)amino]ethyl]amino]acetic acid
SMILES C(CN(CC(=O)O)CC(=O)O)N(CCN(CC(=O)O)CC(=O)O)CC(=O)O

Specifications

Melting Point 220.0°C
Color White
Density 1.5600g/mL
Flash Point 200°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Plastic bottle
Quantity 250 g
Beilstein 04,III,119
Merck Index 15,7237
Specific Gravity 1.56
Solubility Information Solubility in water: 5g/L (20°C), 13g/L (50°C), 48g/L (80°C). Other solubilities: soluble in alkali
Formula Weight 393.34
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material Diethylenetriaminepentaacetic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-652-8

RUO – Research Use Only