missing translation for 'onlineSavingsMsg'
Learn More

Diethylenetriaminepentaacetic Acid, 98%, Spectrum™ Chemical

Catalog No. 18610196
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610196 100 g Poly Bottle
18610197 500 g Poly Bottle
18610198 2.5 kg Poly Bottle
18610199 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18610196 Supplier Spectrum Chemical Mfg Cor Supplier No. D2323100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 67-43-6

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 100 g
Percent Purity 0.98
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.