missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylene Glycol Monohexyl Ether 98.0+%, TCI America™
$29.83 - $57.79
Chemical Identifiers
| CAS | 112-59-4 |
|---|---|
| Molecular Formula | C10H22O3 |
| Molecular Weight (g/mol) | 190.28 |
| MDL Number | MFCD00010703 |
| InChI Key | GZMAAYIALGURDQ-UHFFFAOYSA-N |
| Synonym | diethylene glycol monohexyl ether, 2-2-hexyloxyethoxy ethanol, hexol carbitol, hexyl carbitol, n-hexyl carbitol, n-hexoxyethoxyethanol, 3,6-dioxadodecanol-1, ethanol, 2-2-hexyloxy ethoxy, hexyldiglycol, hexylkarbitol |
| PubChem CID | 8199 |
| IUPAC Name | 2-[2-(hexyloxy)ethoxy]ethan-1-ol |
| SMILES | CCCCCCOCCOCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D050125ML
|
TCI America
D050125ML |
25 mL |
Each for $29.83
|
|
|||||
|
D0501500ML
|
TCI America
D0501500ML |
500 mL |
Each for $57.79
|
|
|||||
Chemical Identifiers
| 112-59-4 | |
| 190.28 | |
| GZMAAYIALGURDQ-UHFFFAOYSA-N | |
| 8199 | |
| CCCCCCOCCOCCO |
| C10H22O3 | |
| MFCD00010703 | |
| diethylene glycol monohexyl ether, 2-2-hexyloxyethoxy ethanol, hexol carbitol, hexyl carbitol, n-hexyl carbitol, n-hexoxyethoxyethanol, 3,6-dioxadodecanol-1, ethanol, 2-2-hexyloxy ethoxy, hexyldiglycol, hexylkarbitol | |
| 2-[2-(hexyloxy)ethoxy]ethan-1-ol |
Specifications
| 112-59-4 | |
| 258°C | |
| MFCD00010703 | |
| diethylene glycol monohexyl ether, 2-2-hexyloxyethoxy ethanol, hexol carbitol, hexyl carbitol, n-hexyl carbitol, n-hexoxyethoxyethanol, 3,6-dioxadodecanol-1, ethanol, 2-2-hexyloxy ethoxy, hexyldiglycol, hexylkarbitol | |
| CCCCCCOCCOCCO | |
| 190.28 | |
| 190.28 | |
| Liquid |
| Yellow | |
| C10H22O3 | |
| 25 mL | |
| GZMAAYIALGURDQ-UHFFFAOYSA-N | |
| 2-[2-(hexyloxy)ethoxy]ethan-1-ol | |
| 8199 | |
| ≥98.0% (GC) | |
| Diethylene Glycol Monohexyl Ether |
Safety and Handling
EINECSNumber : (2)-2424
RTECSNumber : KL2625000
TSCA : Yes