missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol Monohexyl Ether 98.0+%, TCI America™

Catalog No. D0501500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0501500ML 500 mL
D050125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0501500ML Supplier TCI America Supplier No. D0501500ML

Chemical Identifiers

CAS 112-59-4
Molecular Formula C10H22O3
Molecular Weight (g/mol) 190.28
MDL Number MFCD00010703
InChI Key GZMAAYIALGURDQ-UHFFFAOYSA-N
Synonym diethylene glycol monohexyl ether, 2-2-hexyloxyethoxy ethanol, hexol carbitol, hexyl carbitol, n-hexyl carbitol, n-hexoxyethoxyethanol, 3,6-dioxadodecanol-1, ethanol, 2-2-hexyloxy ethoxy, hexyldiglycol, hexylkarbitol
PubChem CID 8199
IUPAC Name 2-[2-(hexyloxy)ethoxy]ethan-1-ol
SMILES CCCCCCOCCOCCO

Specifications

Color Yellow
Boiling Point 258°C
Quantity 500 mL
Formula Weight 190.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethylene Glycol Monohexyl Ether
EINECSNumber (2)-2424
RTECSNumber KL2625000
TSCA Yes