missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol Monoethyl Ether Acetate 99.0+%, TCI America™

Catalog No. D050025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D050025G 25 g
D0500500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D050025G Supplier TCI America Supplier No. D050025G

Chemical Identifiers

CAS 112-15-2
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.21
MDL Number MFCD00041928
InChI Key FPZWZCWUIYYYBU-UHFFFAOYSA-N
Synonym carbitol acetate, 2-2-ethoxyethoxy ethyl acetate, diethylene glycol monoethyl ether acetate, ektasolve de acetate, karbitolacetat, ethoxydiglycol acetate, glycol ether de acetate, diglycol monoethyl ether acetate, ethanol, 2-2-ethoxyethoxy-, acetate, ethyldiglycol acetate
PubChem CID 8165
IUPAC Name 2-(2-ethoxyethoxy)ethyl acetate
SMILES CCOCCOCCOC(C)=O

Specifications

Color Colorless
Boiling Point 218°C
Quantity 25 g
Formula Weight 176.21
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethylene Glycol Monoethyl Ether Acetate
EINECSNumber (2)-0744
RTECSNumber KK8925000
TSCA Yes