missing translation for 'onlineSavingsMsg'
Learn More

Diethylene glycol monoethyl ether acetate, 99%

Catalog No. p-7046205
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL13446AE 100 mL
AAL13446AP 500 mL
AAL134460F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL13446AE Supplier Thermo Scientific Chemicals Supplier No. L13446AE
Only null left

CAS: 112-15-2 | C8H16O4 | 176.21 g/mol

Diethylene glycol monoethyl ether acetate is used as a catalytic agent, petrochemical additive, coating auxiliary agent and in organic synthesis. It is mainly used as metal, furniture paint solvents and as organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylene glycol monoethyl ether acetate is used as a catalytic agent, petrochemical additive, coating auxiliary agent and in organic synthesis. It is mainly used as metal, furniture paint solvents and as organic intermediate.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with bases and oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Containers which are opened must be carefully resealed and kept upright to prevent leakage.

Chemical Identifiers

CAS 112-15-2
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.21
MDL Number MFCD00041928
InChI Key FPZWZCWUIYYYBU-UHFFFAOYSA-N
Synonym carbitol acetate, 2-2-ethoxyethoxy ethyl acetate, diethylene glycol monoethyl ether acetate, ektasolve de acetate, karbitolacetat, ethoxydiglycol acetate, glycol ether de acetate, diglycol monoethyl ether acetate, ethanol, 2-2-ethoxyethoxy-, acetate, ethyldiglycol acetate
PubChem CID 8165
IUPAC Name 2-(2-ethoxyethoxy)ethyl acetate
SMILES CCOCCOCCOC(C)=O

Specifications

Melting Point -25°C
Density 1.012
Boiling Point 218°C to 219°C
Flash Point 95°C (203°F)
Refractive Index 1.421
Quantity 100 mL
Beilstein 1764643
Merck Index 14,1800
Solubility Information Soluble in water.
Formula Weight 176.21
Percent Purity 99%
Chemical Name or Material Diethylene glycol monoethyl ether acetate
Show More Show Less
Hazard Category H319-H335
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
EINECSNumber 203-940-1
RTECSNumber KK8925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only