missing translation for 'onlineSavingsMsg'
Learn More

Diethylene glycol monoethyl ether, 98%

Catalog No. p-7022068
Change view
Click to view available options
Quantity:
10,000 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA146700C 10,000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA146700C Supplier Thermo Scientific Chemicals Supplier No. A146700C
Only null left

CAS: 111-90-0 | C6H14O3 | 134.175 g/mol

Diethylene glycol monoethyl ether has low setting point and low viscocity at low temperature so it is used in manufacturing brake fluid. It is used as a flow and gloss promoter in paint industries, in production of printing ink and as a cleaner in offset printing. Also used in textile as a solvent for dyestuff in the printing and dying of fiber & fabrics, in the production & wood preservaties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylene glycol monoethyl ether has low setting point and low viscocity at low temperature so it is used in manufacturing brake fluid. It is used as a flow and gloss promoter in paint industries, in production of printing ink and as a cleaner in offset printing. Also used in textile as a solvent for dyestuff in the printing and dying of fiber & fabrics, in the production & wood preservaties.

Solubility
Fully miscible in water.

Notes
It is hygroscopic. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 111-90-0
Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.175
MDL Number MFCD00002872
InChI Key XXJWXESWEXIICW-UHFFFAOYSA-N
Synonym diethylene glycol monoethyl ether, 2-2-ethoxyethoxy ethanol, carbitol, transcutol, ethoxy diglycol, dioxitol, ethyl carbitol, ethyl digol, carbitol solvent, ethanol, 2-2-ethoxyethoxy
PubChem CID 8146
ChEBI CHEBI:40572
IUPAC Name 2-(2-ethoxyethoxy)ethanol
SMILES CCOCCOCCO

Specifications

Melting Point -80°C
Density 0.989
Boiling Point 201°C to 202°C
Flash Point 96°C (204°F)
Odor Pleasant
Refractive Index 1.428
Linear Formula HOCH2CH2OCH2CH2OCH2CH3
Quantity 10,000 g
Beilstein 1736441
Sensitivity Hygroscopic
Merck Index 14,1800
Solubility Information Fully miscible in water.
Formula Weight 134.18
Percent Purity 98%
Chemical Name or Material Diethylene glycol monoethyl ether
Show More Show Less
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 203-919-7
RTECSNumber KK8750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only