missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol Monobutyl Ether 99.0+%, TCI America™

Catalog No. p-7118617
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B069925ML 25 mL
B0699500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B069925ML Supplier TCI America Supplier No. B069925ML
Only null left

Chemical Identifiers

CAS 112-34-5
Molecular Formula C8H18O3
Molecular Weight (g/mol) 162.229
MDL Number MFCD00002881
InChI Key OAYXUHPQHDHDDZ-UHFFFAOYSA-N
Synonym 2-2-butoxyethoxy ethanol, diethylene glycol monobutyl ether, butyldiglycol, butyl carbitol, diethylene glycol butyl ether, butyl diglycol, butyl digol, ethanol, 2-2-butoxyethoxy, butoxyethoxyethanol, butoxydiglycol
PubChem CID 8177
IUPAC Name 2-(2-butoxyethoxy)ethanol
SMILES CCCCOCCOCCO

Specifications

Melting Point -68°C
Color Colorless
Boiling Point 230°C
Quantity 25 mL
Formula Weight 162.23
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethylene Glycol Monobutyl Ether
EINECSNumber (2)-0422&(7)-0097
RTECSNumber KJ9100000
TSCA Yes