missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diéthylène glycol Monobutylique Éther 99,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | -68°C |
| Color | Incolore |
| Boiling Point | 230°C |
| Quantity | 25 mL |
| Formula Weight | 162.23 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | Diethylene Glycol Monobutyl Ether |
Sécurité et manipulation
| EINECSNumber | (2)-0422&(7)-0097 |
| RTECSNumber | KJ9100000 |
| TSCA | Yes |