missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol Monobutyl Ether Acetate 98.0+%, TCI America™

Catalog No. p-7126250
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
D049925ML 25 mL
D0499500ML 500 mL
2 options

Catalog No. D049925ML

Supplier: TCI America D049925ML

Only null left

Chemical Identifiers

CAS 124-17-4
Molecular Formula C10H20O4
Molecular Weight (g/mol) 204.266
MDL Number MFCD00009458
InChI Key VXQBJTKSVGFQOL-UHFFFAOYSA-N
Synonym 2-2-butoxyethoxy ethyl acetate, butoxyethoxyethyl acetate, diethylene glycol monobutyl ether acetate, butyl diglycol acetate, butyl carbitol acetate, ektasolve db acetate, 2-2-butoxyethoxy ethanol acetate, butyl diethylene glycol acetate, butylkarbitolacetat, diglycol monobutyl ether acetate
PubChem CID 31288
IUPAC Name 2-(2-butoxyethoxy)ethyl acetate
SMILES CCCCOCCOCCOC(=O)C

Specifications

Melting Point -32°C
Color Colorless
Boiling Point 247°C
Quantity 25 mL
Formula Weight 204.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethylene Glycol Monobutyl Ether Acetate
EINECSNumber (2)-0744
RTECSNumber KJ9275000
TSCA Yes