missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol Monobutyl Ether Acetate 98.0+%, TCI America™

Catalog No. D0499500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0499500ML 500 mL
D049925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0499500ML Supplier TCI America Supplier No. D0499500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 124-17-4
Molecular Formula C10H20O4
Molecular Weight (g/mol) 204.266
MDL Number MFCD00009458
InChI Key VXQBJTKSVGFQOL-UHFFFAOYSA-N
Synonym 2-2-butoxyethoxy ethyl acetate, butoxyethoxyethyl acetate, diethylene glycol monobutyl ether acetate, butyl diglycol acetate, butyl carbitol acetate, ektasolve db acetate, 2-2-butoxyethoxy ethanol acetate, butyl diethylene glycol acetate, butylkarbitolacetat, diglycol monobutyl ether acetate
PubChem CID 31288
IUPAC Name 2-(2-butoxyethoxy)ethyl acetate
SMILES CCCCOCCOCCOC(=O)C

Specifications

Melting Point -32°C
Color Colorless
Boiling Point 247°C
Quantity 500 mL
Formula Weight 204.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethylene Glycol Monobutyl Ether Acetate
EINECSNumber (2)-0744
RTECSNumber KJ9275000
TSCA Yes