missing translation for 'onlineSavingsMsg'
Learn More

Diethylene glycol mono-n-hexyl ether, 97%

Catalog No. AAB20774AP
Change view
Click to view available options
Quantity:
10,000 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB20774AP 500 mL
AAB207740F 2500 mL
AAB207740D 10,000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB20774AP Supplier Thermo Scientific Chemicals Supplier No. B20774AP

CAS: 112-59-4 | C10H22O3 | 190.28 g/mol

Used as a solvent for coatings and degreasing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a solvent for coatings and degreasing.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 112-59-4
Molecular Formula C10H22O3
Molecular Weight (g/mol) 190.28
MDL Number MFCD00010703
InChI Key GZMAAYIALGURDQ-UHFFFAOYSA-N
Synonym diethylene glycol monohexyl ether, 2-2-hexyloxyethoxy ethanol, hexol carbitol, hexyl carbitol, n-hexyl carbitol, n-hexoxyethoxyethanol, 3,6-dioxadodecanol-1, ethanol, 2-2-hexyloxy ethoxy, hexyldiglycol, hexylkarbitol
PubChem CID 8199
IUPAC Name 2-(2-hexoxyethoxy)ethanol
SMILES CCCCCCOCCOCCO

Specifications

Melting Point -40°C
Density 0.932
Boiling Point 259°C to 261°C
Flash Point 123°C (253°F)
Refractive Index 1.437
Linear Formula HOCH2CH2OCH2CH2O(CH2)5CH3
Quantity 500 mL
Beilstein 1743959
Solubility Information Soluble in water.
Formula Weight 190.28
Percent Purity 97%
Chemical Name or Material Diethylene glycol mono-n-hexyl ether
Show More Show Less
Hazard Category H312-H318
Hazard Statement GHS H Statement
H318-H312
Causes serious eye damage.
Harmful in contact with skin.
Precautionary Statement P280-P302+P352-P305+P351+P338-P310-P312-P363-P501c
EINECSNumber 203-988-3
RTECSNumber KL2625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only