missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol Ethyl Methyl Ether 98.0+%, TCI America™

Catalog No. D4280500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4280500ML 500 mL
D428025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4280500ML Supplier TCI America Supplier No. D4280500ML
Only null left

Chemical Identifiers

CAS 1002-67-1
Molecular Formula C7H16O3
Molecular Weight (g/mol) 148.20
MDL Number MFCD01727263
InChI Key CNJRPYFBORAQAU-UHFFFAOYSA-N
Synonym 2,5,8-Trioxadecane, 1-Ethoxy-2-(2-methoxyethoxy)ethane
PubChem CID 13847
IUPAC Name 1-ethoxy-2-(2-methoxyethoxy)ethane
SMILES CCOCCOCCOC

Specifications

Color Colorless
Boiling Point 179°C
Quantity 500 mL
UN Number 3271
Formula Weight 148.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethylene Glycol Ethyl Methyl Ether
EINECSNumber (7)-1321
RTECSNumber KN9650000
TSCA Yes