missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylene Glycol Diethyl Ether 98.0+%, TCI America™
Supplier: TCI America B048925ML
Specifications
| Diethylene Glycol Diethyl Ether | |
| -44°C | |
| 188°C | |
| MFCD00009254 | |
| diethylene glycol diethyl ether, 2-ethoxyethyl ether, diethyl carbitol, ethyl diglyme, 1-ethoxy-2-2-ethoxyethoxy ethane, bis 2-ethoxyethyl ether, diethyldiethylene glycol, 3,6,9-trioxaundecane, degdee, ether, bis 2-ethoxyethyl | |
| CCOCCOCCOCC | |
| 162.229 | |
| CHEBI:44664 | |
| ≥98.0% (GC) |
| 112-36-7 | |
| Colorless | |
| C8H18O3 | |
| 25 mL | |
| RRQYJINTUHWNHW-UHFFFAOYSA-N | |
| 1-ethoxy-2-(2-ethoxyethoxy)ethane | |
| 8179 | |
| 162.23 | |
| Liquid |
Chemical Identifiers
| 112-36-7 | |
| 162.229 | |
| RRQYJINTUHWNHW-UHFFFAOYSA-N | |
| 8179 | |
| 1-ethoxy-2-(2-ethoxyethoxy)ethane |
| C8H18O3 | |
| MFCD00009254 | |
| diethylene glycol diethyl ether, 2-ethoxyethyl ether, diethyl carbitol, ethyl diglyme, 1-ethoxy-2-2-ethoxyethoxy ethane, bis 2-ethoxyethyl ether, diethyldiethylene glycol, 3,6,9-trioxaundecane, degdee, ether, bis 2-ethoxyethyl | |
| CHEBI:44664 | |
| CCOCCOCCOCC |
Safety and Handling
EINECSNumber : (2)-0433&(7)-1321
RTECSNumber : KN3160000
TSCA : Yes