missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylene Glycol Dibutyl Ether 98.0+%, TCI America™
$55.92 - $214.37
Chemical Identifiers
| CAS | 112-73-2 |
|---|---|
| Molecular Formula | C12H26O3 |
| Molecular Weight (g/mol) | 218.337 |
| MDL Number | MFCD00009459 |
| InChI Key | KZVBBTZJMSWGTK-UHFFFAOYSA-N |
| Synonym | diethylene glycol dibutyl ether, bis 2-butoxyethyl ether, dibutyl carbitol, butyl diglyme, 1-2-2-butoxyethoxy ethoxy butane, 5,8,11-trioxapentadecane, ether, bis butoxyethyl, 2,2'-dibutoxyethyl ether, ether, bis 2-butoxyethyl |
| PubChem CID | 8210 |
| IUPAC Name | 1-[2-(2-butoxyethoxy)ethoxy]butane |
| SMILES | CCCCOCCOCCOCCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B082825ML
|
TCI America
B082825ML |
25 mL |
Each for $55.92
|
|
|||||
|
B0828500ML
|
TCI America
B0828500ML |
500 mL |
Each for $214.37
|
|
|||||
Chemical Identifiers
| 112-73-2 | |
| 218.337 | |
| KZVBBTZJMSWGTK-UHFFFAOYSA-N | |
| 8210 | |
| CCCCOCCOCCOCCCC |
| C12H26O3 | |
| MFCD00009459 | |
| diethylene glycol dibutyl ether, bis 2-butoxyethyl ether, dibutyl carbitol, butyl diglyme, 1-2-2-butoxyethoxy ethoxy butane, 5,8,11-trioxapentadecane, ether, bis butoxyethyl, 2,2'-dibutoxyethyl ether, ether, bis 2-butoxyethyl | |
| 1-[2-(2-butoxyethoxy)ethoxy]butane |
Specifications
| 112-73-2 | |
| Colorless | |
| C12H26O3 | |
| 25 mL | |
| KZVBBTZJMSWGTK-UHFFFAOYSA-N | |
| 1-[2-(2-butoxyethoxy)ethoxy]butane | |
| 8210 | |
| ≥98.0% (GC) | |
| Diethylene Glycol Dibutyl Ether |
| -60°C | |
| 255°C | |
| MFCD00009459 | |
| diethylene glycol dibutyl ether, bis 2-butoxyethyl ether, dibutyl carbitol, butyl diglyme, 1-2-2-butoxyethoxy ethoxy butane, 5,8,11-trioxapentadecane, ether, bis butoxyethyl, 2,2'-dibutoxyethyl ether, ether, bis 2-butoxyethyl | |
| CCCCOCCOCCOCCCC | |
| 218.337 | |
| 218.34 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-3870&(7)-1321
RTECSNumber : KN0350000
TSCA : Yes