missing translation for 'onlineSavingsMsg'
Learn More

Diethylene glycol dibutyl ether, 99+%

Catalog No. AA42150AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42150AP 500 mL
AA42150AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42150AP Supplier Thermo Scientific Chemicals Supplier No. 042150AP

Extraction of precious metals | CAS: 112-73-2 | C12H26O3 | 218.337 g/mol

Extraction of precious metalsDiethylene glycol dibutyl ether is used as a solvent in Grignard reactions. It is also used as solvents in gold refining, decorative inks for ceramics and digital inks. It finds application in electrochemistry, gas absorption, extractant and high boiling reaction medium. It is also used in fuel, lubricant, textile and medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112-73-2
Molecular Formula C12H26O3
Molecular Weight (g/mol) 218.337
MDL Number MFCD00009459
InChI Key KZVBBTZJMSWGTK-UHFFFAOYSA-N
Synonym diethylene glycol dibutyl ether, bis 2-butoxyethyl ether, dibutyl carbitol, butyl diglyme, 1-2-2-butoxyethoxy ethoxy butane, 5,8,11-trioxapentadecane, ether, bis butoxyethyl, 2,2'-dibutoxyethyl ether, ether, bis 2-butoxyethyl
PubChem CID 8210
IUPAC Name 1-[2-(2-butoxyethoxy)ethoxy]butane
SMILES CCCCOCCOCCOCCCC

Specifications

Melting Point -60°C
Color Colorless
Density 0.885 g/mL
Boiling Point 256°C
Flash Point 117°C (243°F)
Refractive Index 1.4235
Quantity 500 mL
Solubility Information Miscible with dimethyl sulfoxide,ethanol and acetone. Immiscible with water.
Formula Weight 218.74
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Diethylene glycol dibutyl ether
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 204-001-9
RTECSNumber KN0350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only