missing translation for 'onlineSavingsMsg'
Learn More

Diethylene glycol dibutyl ether, 99+%, pure

CAS: 112-73-2 | C12H26O3 | 218.34 g/mol

$438.39 - $438.39

Chemical Identifiers

CAS 112-73-2
Molecular Formula C12H26O3
Molecular Weight (g/mol) 218.34
InChI Key KZVBBTZJMSWGTK-UHFFFAOYSA-N
Synonym diethylene glycol dibutyl ether, bis 2-butoxyethyl ether, dibutyl carbitol, butyl diglyme, 1-2-2-butoxyethoxy ethoxy butane, 5,8,11-trioxapentadecane, ether, bis butoxyethyl, 2,2'-dibutoxyethyl ether, ether, bis 2-butoxyethyl
PubChem CID 8210
IUPAC Name 1-[2-(2-butoxyethoxy)ethoxy]butane
SMILES CCCCOCCOCCOCCCC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC194980010
SDS View Documents
Thermo Scientific Chemicals
194980010
1 L Glass bottle
Each for $438.39
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

112-73-2
218.34
diethylene glycol dibutyl ether, bis 2-butoxyethyl ether, dibutyl carbitol, butyl diglyme, 1-2-2-butoxyethoxy ethoxy butane, 5,8,11-trioxapentadecane, ether, bis butoxyethyl, 2,2'-dibutoxyethyl ether, ether, bis 2-butoxyethyl
1-[2-(2-butoxyethoxy)ethoxy]butane
C12H26O3
KZVBBTZJMSWGTK-UHFFFAOYSA-N
8210
CCCCOCCOCCOCCCC

Specifications

112-73-2
0.8800g/mL
118°C
99% min. (GC)
C12H26O3
[CH3(CH2)3OCH2CH2]2O
0.88
Solubility in water: moderate soluble (3g/L at 20°C). Other solubilities: soluble in chloroform
CCCCOCCOCCOCCCC
218.34
2.39 mPa.s (20°C)
99+%
Liquid
-60.0°C
256.0°C
Authentic
Glass bottle
1.4225 to 1.4245
1 L
diethylene glycol dibutyl ether, bis 2-butoxyethyl ether, dibutyl carbitol, butyl diglyme, 1-2-2-butoxyethoxy ethoxy butane, 5,8,11-trioxapentadecane, ether, bis butoxyethyl, 2,2'-dibutoxyethyl ether, ether, bis 2-butoxyethyl
KZVBBTZJMSWGTK-UHFFFAOYSA-N
1-[2-(2-butoxyethoxy)ethoxy]butane
8210
218.34
Pure
Diethylene glycol dibutyl ether
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 204-001-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only