missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol Dibenzoate 97.0+%, TCI America™

Catalog No. D1522500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1522500ML 500 mL
D152225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1522500ML Supplier TCI America Supplier No. D1522500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 120-55-8
Molecular Formula C18H18O5
Molecular Weight (g/mol) 314.34
MDL Number MFCD00020679
InChI Key NXQMCAOPTPLPRL-UHFFFAOYSA-N
PubChem CID 8437
IUPAC Name 2-[2-(benzoyloxy)ethoxy]ethyl benzoate
SMILES O=C(OCCOCCOC(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 236°C
Quantity 500 mL
Formula Weight 314.34
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethylene Glycol Dibenzoate
RTECSNumber ID6650000
TSCA Yes