missing translation for 'onlineSavingsMsg'
Learn More

Diethylene glycol diacrylate, tech. 75%, stab.

Catalog No. AAL1068214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1068214 25 g
AAL1068222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1068214 Supplier Thermo Scientific Chemicals Supplier No. L1068214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4074-88-8 | C10H14O5 | 214.22 g/mol

Diethylene glycol diacrylate is used as reactive diluent and cross linking agent of radiation curing system, and also may be used as crosslinking resins, plastics and rubber modifier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylene glycol diacrylate is used as reactive diluent and cross linking agent of radiation curing system, and also may be used as crosslinking resins, plastics and rubber modifier.

Solubility
Slightly soluble in water (1-5g/100ml at 18°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive to light. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 4074-88-8
Molecular Formula C10H14O5
Molecular Weight (g/mol) 214.22
MDL Number MFCD00014939
InChI Key LEJBBGNFPAFPKQ-UHFFFAOYSA-N
Synonym diethylene glycol diacrylate, oxydiethylene acrylate, oxydiethylene diacrylate, acrylic acid, oxydiethylene ester, tga 2, 2-propenoic acid, oxydi-2,1-ethanediyl ester, diacrylate diethylene glycol, unii-jk01t392ju, acrylic acid, 2-ethoxyethanol diester, di ethylene glycol diacrylate
PubChem CID 19996
IUPAC Name 2-(2-prop-2-enoyloxyethoxy)ethyl prop-2-enoate
SMILES C=CC(=O)OCCOCCOC(=O)C=C

Specifications

Density 1.118
Boiling Point 94°C (0.2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.462
Quantity 25 g
UN Number UN2810
Beilstein 1953711
Solubility Information Slightly soluble in water (1-5g/100ml at 18°C).
Formula Weight 214.22
Percent Purity 75%
Grade Technical
Chemical Name or Material Diethylene glycol diacrylate, stabilized
Show More Show Less
Hazard Category H311-H315-H317-H319
Hazard Statement GHS H Statement
H311-H315-H319-H317
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P272-P280-P302+P352-P305+P351+P338-P312-P333+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 223-791-6
RTECSNumber AS9450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only