missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylene Glycol Diacrylate (stabilized with HQ + MEHQ) 75.0+%, TCI America™
Supplier: TCI America D5381100G
Specifications
| Diethylene Glycol Diacrylate (stabilized with HQ + MEHQ) | |
| Yellow | |
| C10H14O5 | |
| 100 g | |
| diethylene glycol diacrylate, oxydiethylene acrylate, oxydiethylene diacrylate, acrylic acid, oxydiethylene ester, tga 2, 2-propenoic acid, oxydi-2,1-ethanediyl ester, diacrylate diethylene glycol, unii-jk01t392ju, acrylic acid, 2-ethoxyethanol diester, di ethylene glycol diacrylate | |
| C=CC(=O)OCCOCCOC(=O)C=C | |
| 214.22 | |
| 214.22 | |
| Liquid |
| 4074-88-8 | |
| 96°C | |
| MFCD00014939 | |
| 2810 | |
| LEJBBGNFPAFPKQ-UHFFFAOYSA-N | |
| 2-[2-(prop-2-enoyloxy)ethoxy]ethyl prop-2-enoate | |
| 19996 | |
| ≥75.0% (GC) |
Chemical Identifiers
| 4074-88-8 | |
| 214.22 | |
| LEJBBGNFPAFPKQ-UHFFFAOYSA-N | |
| 19996 | |
| C=CC(=O)OCCOCCOC(=O)C=C |
| C10H14O5 | |
| MFCD00014939 | |
| diethylene glycol diacrylate, oxydiethylene acrylate, oxydiethylene diacrylate, acrylic acid, oxydiethylene ester, tga 2, 2-propenoic acid, oxydi-2,1-ethanediyl ester, diacrylate diethylene glycol, unii-jk01t392ju, acrylic acid, 2-ethoxyethanol diester, di ethylene glycol diacrylate | |
| 2-[2-(prop-2-enoyloxy)ethoxy]ethyl prop-2-enoate |
Safety and Handling
EINECSNumber : (7)-0152
RTECSNumber : AS9450000
TSCA : Yes