missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol Diacrylate (stabilized with HQ + MEHQ) 75.0+%, TCI America™

Catalog No. D538125G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D538125G 25 g
D5381100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D538125G Supplier TCI America Supplier No. D538125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4074-88-8
Molecular Formula C10H14O5
Molecular Weight (g/mol) 214.22
MDL Number MFCD00014939
InChI Key LEJBBGNFPAFPKQ-UHFFFAOYSA-N
Synonym diethylene glycol diacrylate, oxydiethylene acrylate, oxydiethylene diacrylate, acrylic acid, oxydiethylene ester, tga 2, 2-propenoic acid, oxydi-2,1-ethanediyl ester, diacrylate diethylene glycol, unii-jk01t392ju, acrylic acid, 2-ethoxyethanol diester, di ethylene glycol diacrylate
PubChem CID 19996
IUPAC Name 2-[2-(prop-2-enoyloxy)ethoxy]ethyl prop-2-enoate
SMILES C=CC(=O)OCCOCCOC(=O)C=C

Specifications

Color Yellow
Boiling Point 96°C
Quantity 25 g
UN Number 2810
Formula Weight 214.22
Percent Purity ≥75.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethylene Glycol Diacrylate (stabilized with HQ + MEHQ)
EINECSNumber (7)-0152
RTECSNumber AS9450000
TSCA Yes