missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Diethylene Glycol
Description
Diethylene Glycol
Specifications
Specifications
| CAS | 524-38-9 |
| Melting Point | 233°C |
| Molecular Formula | C8H5NO3 |
| Linear Formula | C8H5NO3 |
| Refractive Index | n20/D 1.447 |
| MDL Number | MFCD00005891 |
| Quantity | 500 g |
| Synonym | 2,2'-Oxydiethanol; 2-Hydroxyethyl ether; Bis(2-hydroxyethyl) ether; Diglycol |
| Molecular Weight (g/mol) | 163.13 g/mol |