missing translation for 'onlineSavingsMsg'
Learn More

Diethyle3,4-dihydroxyfuran-2,5-dicarboxylate, 97%, Thermo Scientific™

Catalog No. BTB10007CB
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB10007CB 250mg Amber Glass Bottle
BTB10007DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTB10007CB Supplier Thermo Scientific Chemicals Supplier No. BTB10007CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6270-57-1
Molecular Formula C10H12O7
Molecular Weight (g/mol) 244.199
MDL Number MFCD00092386
InChI Key JGCMPUKYPRCPRE-UHFFFAOYSA-N
Synonym 2,5-diethyl 3,4-dihydroxyfuran-2,5-dicarboxylate, diethyl 3,4-dihydroxy-2,5-furandicarboxylate, 2,5-bis ethoxy hydroxy methylidene oxolane-3,4-dione, 3,4-dihydroxy-furan-2,5-dicarboxylic acid diethyl ester, pubchem7048, maybridge4_000491, 2,5-bis ethoxycarbonyl-3,4-dihydroxyfuran, 2,5-bis ethoxy oxidanyl methylidene oxolane-3,4-dione, 2z,5z-bis ethoxy hydroxy methylidene oxolane-3,4-dione, 2e,5e-2,5-bis ethoxy hydroxy methylidene oxolane-3,4-dione
PubChem CID 234848
IUPAC Name diethyl 3,4-dihydroxyfuran-2,5-dicarboxylate
SMILES CCOC(=O)C1=C(C(=C(O1)C(=O)OCC)O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 244.2
Percent Purity 97%
Chemical Name or Material Diethyl 3, 4-dihydroxyfuran-2, 5-dicarboxylate