missing translation for 'onlineSavingsMsg'
Learn More

Diethyldithiocarbamic acid, silver salt, ACS reagent

Catalog No. AC419330250
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC419330250 25 g Glass bottle
AC419330100 10 g Glass bottle
AC419331000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC419330250 Supplier Thermo Scientific Chemicals Supplier No. 419330250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1470-61-7 | C5H10AgNS2 | 256.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1470-61-7
Molecular Formula C5H10AgNS2
Molecular Weight (g/mol) 256.13
MDL Number MFCD00004929
InChI Key NSVHDIYWJVLAGH-UHFFFAOYSA-M
Synonym silver diethyldithiocarbamate, unii-gvn001b86o, agddtc, silver n,n-diethylcarbamodithioate, diethyldithiocarbamic acid, silver salt, diethyl dithio carbamic acid silver salt, silver i diethyldithiocarbamate, silver, diethylcarbamodithioato-ks,ks', ditiocarb silver, silver n,n-diethyldithiocarbamate
PubChem CID 3034078
IUPAC Name silver;N,N-diethylcarbamodithioate
SMILES CCN(CC)C(=S)[S-].[Ag+]

Specifications

Melting Point 172.0°C to 175.0°C
Color Khaki
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula (C2H5)2NCS2Ag
Quantity 25 g
Beilstein 04,121
Solubility Information (in Pyridine) Passes test
Formula Weight 256.13
Grade ACS Reagent
Physical Form Amorphous Powder
Chemical Name or Material Diethyldithiocarbamic acid, silver salt
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective glov
EINECSNumber 216-003-7
TSCA TSCA

RUO – Research Use Only