Learn More
Diethyldithiocarbamic acid, silver salt, 99%, for analysis
CAS: 1470-61-7 | C5H10AgNS2 | 256.13 g/mol
Supplier: Thermo Scientific Chemicals 114290250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysisSpecifications
| Diethyldithiocarbamic acid, silver salt | |
| 1470-61-7 | |
| Green to Khaki | |
| 99% | |
| C5H10AgNS2 | |
| MFCD00004929 | |
| 04, 121 | |
| (0.5% in pyridine) clear | |
| CCN(CC)C(=S)[S-].[Ag+] | |
| 256.13 | |
| 256.13 | |
| Analysis |
| For analysis | |
| 172.0°C to 175.0°C | |
| Authentic | |
| Glass bottle | |
| (C2H5)2NCS2Ag | |
| 25 g | |
| silver diethyldithiocarbamate, unii-gvn001b86o, agddtc, silver n,n-diethylcarbamodithioate, diethyldithiocarbamic acid, silver salt, diethyl dithio carbamic acid silver salt, silver i diethyldithiocarbamate, silver, diethylcarbamodithioato-ks,ks', ditiocarb silver, silver n,n-diethyldithiocarbamate | |
| NSVHDIYWJVLAGH-UHFFFAOYSA-M | |
| silver;N,N-diethylcarbamodithioate | |
| 3034078 | |
| 99% | |
| Amorphous Powder |
Chemical Identifiers
| 1470-61-7 | |
| 256.13 | |
| NSVHDIYWJVLAGH-UHFFFAOYSA-M | |
| 3034078 | |
| CCN(CC)C(=S)[S-].[Ag+] |
| C5H10AgNS2 | |
| MFCD00004929 | |
| silver diethyldithiocarbamate, unii-gvn001b86o, agddtc, silver n,n-diethylcarbamodithioate, diethyldithiocarbamic acid, silver salt, diethyl dithio carbamic acid silver salt, silver i diethyldithiocarbamate, silver, diethylcarbamodithioato-ks,ks', ditiocarb silver, silver n,n-diethyldithiocarbamate | |
| silver;N,N-diethylcarbamodithioate |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective glov
GHS Signal Word: Warning
EINECSNumber : 216-003-7
RUO – Research Use Only