missing translation for 'onlineSavingsMsg'
Learn More

Diethylcyanamide 98.0+%, TCI America™

Catalog No. D49651G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49651G 1 g
D49655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49651G Supplier TCI America Supplier No. D49651G

Chemical Identifiers

CAS 617-83-4
Molecular Formula C5H10N2
Molecular Weight (g/mol) 98.15
MDL Number MFCD00001769
InChI Key ZZTSQZQUWBFTAT-UHFFFAOYSA-N
Synonym n,n-diethylcyanamide, cyanamide, diethyl, n-cyanodiethylamine, diethylkyanamid, usaf ek-6326, diethylkyanamid czech, cyanodiethylamine, diethylamino carbonitrile, diethyl-cyanamide, aminodiethylcarbonitrile
PubChem CID 12050
IUPAC Name cyanodiethylamine
SMILES CCN(CC)C#N

Specifications

Color Yellow
Boiling Point 188°C
Quantity 1 g
UN Number 3276
Formula Weight 98.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethylcyanamide
RTECSNumber GS6300000
TSCA No