missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylcarbamoyl Chloride 98.0+%, TCI America™
$61.51 - $285.21
Chemical Identifiers
| CAS | 88-10-8 |
|---|---|
| Molecular Formula | C5H10ClNO |
| Molecular Weight (g/mol) | 135.591 |
| MDL Number | MFCD00000636 |
| InChI Key | OFCCYDUUBNUJIB-UHFFFAOYSA-N |
| Synonym | diethylcarbamoyl chloride, diethylcarbamyl chloride, carbamic chloride, diethyl, diethylcarbamic chloride, carbamoyl chloride, diethyl, diethylchloroformamide, carbamidoyl chloride, diethyl, n,n-diethylchloroformamide, unii-5riv1u6h5w, n,n-diethylcarbamyl chloride |
| PubChem CID | 6916 |
| IUPAC Name | N,N-diethylcarbamoyl chloride |
| SMILES | CCN(CC)C(=O)Cl |
Chemical Identifiers
| 88-10-8 | |
| 135.591 | |
| OFCCYDUUBNUJIB-UHFFFAOYSA-N | |
| 6916 | |
| CCN(CC)C(=O)Cl |
| C5H10ClNO | |
| MFCD00000636 | |
| diethylcarbamoyl chloride, diethylcarbamyl chloride, carbamic chloride, diethyl, diethylcarbamic chloride, carbamoyl chloride, diethyl, diethylchloroformamide, carbamidoyl chloride, diethyl, n,n-diethylchloroformamide, unii-5riv1u6h5w, n,n-diethylcarbamyl chloride | |
| N,N-diethylcarbamoyl chloride |
Specifications
| 88-10-8 | |
| Yellow | |
| MFCD00000636 | |
| OFCCYDUUBNUJIB-UHFFFAOYSA-N | |
| N,N-diethylcarbamoyl chloride | |
| 135.591 | |
| 135.59 | |
| Liquid |
| 195°C | |
| C5H10ClNO | |
| diethylcarbamoyl chloride, diethylcarbamyl chloride, carbamic chloride, diethyl, diethylcarbamic chloride, carbamoyl chloride, diethyl, diethylchloroformamide, carbamidoyl chloride, diethyl, n,n-diethylchloroformamide, unii-5riv1u6h5w, n,n-diethylcarbamyl chloride | |
| CCN(CC)C(=O)Cl | |
| 25 g | |
| 6916 | |
| ≥98.0% (GC,T) | |
| Diethylcarbamoyl Chloride |
Safety and Handling
EINECSNumber : (2)-2859
RTECSNumber : FD4025000
TSCA : Yes