missing translation for 'onlineSavingsMsg'
Learn More

Diethylamine Hydroiodide 97.0+%, TCI America™

Catalog No. D46431G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46431G 1 g
D46435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46431G Supplier TCI America Supplier No. D46431G
Only null left

Chemical Identifiers

CAS 19833-78-4
Molecular Formula C4H12IN
Molecular Weight (g/mol) 201.05
MDL Number MFCD28384142
InChI Key YYMLRIWBISZOMT-UHFFFAOYSA-N
Synonym Diethylammonium Iodide
PubChem CID 88320434
IUPAC Name diethylazanium iodide
SMILES [I-].CC[NH2+]CC

Specifications

Melting Point 175°C
Color White
Quantity 1 g
Formula Weight 201.05
Percent Purity ≥97.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Diethylamine Hydroiodide