missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylamine Hydrochloride 98.5+%, TCI America™
Specifications
Specifications
| Melting Point | 231°C |
| Color | White |
| Boiling Point | 330°C |
| Quantity | 500 g |
| Formula Weight | 109.60 |
| Percent Purity | ≥98.5% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Diethylamine Hydrochloride |
Safety and Handling
| EINECSNumber | (2)-0135 |
| RTECSNumber | IA3084000 |
| TSCA | Yes |