missing translation for 'onlineSavingsMsg'
Learn More

Diethylamine Hydrochloride 98.5+%, TCI America™

Catalog No. D046825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D046825G 25 g
D0468500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D046825G Supplier TCI America Supplier No. D046825G
Only null left

Chemical Identifiers

CAS 660-68-4
Molecular Formula C4H12ClN
Molecular Weight (g/mol) 109.597
MDL Number MFCD00012499
InChI Key HDITUCONWLWUJR-UHFFFAOYSA-N
Synonym diethylamine hydrochloride, diethylammonium chloride, diethyl amine hydrochloride, n-ethylethanamine hydrochloride, ethanamine, n-ethyl-, hydrochloride, unii-ze9v3g1135, ethanamine, n-ethyl-, hydrochloride 1:1, diethylaminehydrochloride, diethylamine hcl, diethyl amine hcl
PubChem CID 10197650
IUPAC Name N-ethylethanamine;hydrochloride
SMILES CCNCC.Cl

Specifications

Melting Point 231°C
Color White
Boiling Point 330°C
Quantity 25 g
Formula Weight 109.60
Percent Purity ≥98.5% (T)
Physical Form Crystalline Powder
Chemical Name or Material Diethylamine Hydrochloride
EINECSNumber (2)-0135
RTECSNumber IA3084000
TSCA Yes