missing translation for 'onlineSavingsMsg'
Learn More

Diethylamine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog No. AAA1363730
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1363730 250 g
AAA136370B 1000 g
AAA136370I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1363730 Supplier Thermo Scientific Chemicals Supplier No. A1363730
Only null left

Diethylamine hydrochloride acts as a precursor of atrazine and lysergic acid diethylamide. It is also used as a reactant in the production of dyes and pharmaceutical compounds such as ranitidine. Further, it is used in Mannich reaction with paraformaldehyde. In addition to this, it is employed in the synthesis of diethylaminoethyl (DEAE) cottons by reacting with cotton cellulose in the presence of sodium hydroxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylamine hydrochloride acts as a precursor of atrazine and lysergic acid diethylamide. It is also used as a reactant in the production of dyes and pharmaceutical compounds such as ranitidine. Further, it is used in Mannich reaction with paraformaldehyde. In addition to this, it is employed in the synthesis of diethylaminoethyl (DEAE) cottons by reacting with cotton cellulose in the presence of sodium hydroxide.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 660-68-4
Molecular Formula C4H12ClN
Molecular Weight (g/mol) 109.597
MDL Number MFCD00012499
InChI Key HDITUCONWLWUJR-UHFFFAOYSA-N
Synonym diethylamine hydrochloride, diethylammonium chloride, diethyl amine hydrochloride, n-ethylethanamine hydrochloride, ethanamine, n-ethyl-, hydrochloride, unii-ze9v3g1135, ethanamine, n-ethyl-, hydrochloride 1:1, diethylaminehydrochloride, diethylamine hcl, diethyl amine hcl
PubChem CID 10197650
IUPAC Name N-ethylethanamine;hydrochloride
SMILES CCNCC.Cl

Specifications

Melting Point 227°C to 230°C
Boiling Point 320°C to 330°C
Quantity 250 g
Beilstein 3590084
Sensitivity Hygroscopic
Merck Index 14,3111
Solubility Information Soluble in water.
Formula Weight 109.6
Percent Purity 99%
Chemical Name or Material Diethylamine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-541-9
RTECSNumber IA3084000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only